3,4-Diethoxy-3-cyclobuten-1,2-dion ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98%
CAS No.: 5231-87-8
Molecular Formula: C8H10O4
Formula Weight: 170.16
Storage Temperature: 2°C to 8°C
Flash Point:
Formel:
C₈H₁₀O₄ Siedepunkt: 95 °C (0,1 mmHg) Dichte: 1,162 Flammpunkt: >110 °C (230 °F) |
MDL:
MFCD00001333 CAS-Nummer: 5231-87-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...