cis-4-Hexen-1-ol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A19266.06
A19266.14
A19266.06EA
37
EUR
A19266.06
A19266.14
cis-4-Hexen-1-ol ≥97%
cis-4-Hexen-1-ol
Purity: 97%
CAS No.: 928-91-6
Molecular Formula: C6H12O
Formula Weight: 100.16
Flash Point: 65°(149°F)
Odour: Pleasant
Boiling Point: 158°
UN No.: 1987
MDL No.: MFCD00148974
Formel:
C₆H₁₂O Siedepunkt: 158 °C Dichte: 0,857 Flammpunkt: 65 °C (149 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00148974 CAS-Nummer: 928-91-6 EINECS: 213-187-0 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...