3,4,5-Trihydroxybenzaldehyd, Hyrdrat 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Gallic aldehyde, Pyrogallol-5-carboxaldehyde

A19270.03 A19270.06
A19270.03EA 113 EUR
A19270.03 A19270.06
3,4,5-Trihydroxybenzaldehyd, Hyrdrat 97%
3,4,5-Trihydroxybenzaldehyde

3,4,5-trihydroxybenzaldehyde from Geum japonicum has dual inhibitory effect on matrix metalloproteinase 9; inhibition of gelatinoytic activity as well as MMP-9 expression in TNF-α induced HASMC.


Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Formel: C₇H₆O₄·1H₂O
Molecular Weight: 154.12 g/mol (anhy)
Schmelzpunkt: ∼ 212 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003371
CAS-Nummer: 207742-88-9
EINECS: 237-168-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR