3-Brom-4-fluorbenzylamin Hydrochlorid 98%

Lieferant: Thermo Scientific
Achtung

A19280.06
A19280.06EA 60.4 EUR
A19280.06
3-Brom-4-fluorbenzylamin Hydrochlorid 98%
3-Brom-4-fluorbenzylamin Hydrochlorid

Purity: 98% CAS No.: 202865-68-7 Molecular Formula: C7H7BrFN.HCl Formula Weight: 240.50 Melting Point: 214-218° MDL No.: MFCD00143094

Formel: C₇H₈BrClFN
Molecular Weight: 240,50 g/mol
Schmelzpunkt: 214…218 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143094
CAS-Nummer: 202865-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR