Chloroform : isoamyl alcohol (24:1) (v/v)

Lieferant: PanReac AppliChem
Achtung

A1935.0500
A1935.0500EA 51.9 EUR
A1935.0500
Chloroform : isoamyl alcohol (24:1) (v/v)
Chloroform : isoamyl alcohol
Formel: CHCl₃,C₅H₁₂O
Lagertemperatur: Raumtemperatur
UN: 1888
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR