5-Brom-o-anisaldehyd ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-Bromo-2-methoxybenzaldehyde, 5-Bromo-2-anisaldehyde
A19396.14
A19396.22
AB125716.0100
A19396.14EA
19.1
EUR
A19396.14
A19396.22
ABCRAB125716.0100
5-Brom-o-anisaldehyd ≥98%
5-Brom-o-anisaldehyd
Purity: 98+%
CAS No.: 25016-01-7
Molecular Formula: C8H7BrO2
Formula Weight: 215.05
Melting Point: 114-118°
Beilstein Reference: 508840
Formel:
C₈H₇BrO₂ Molecular Weight: 215.05 g/mol Siedepunkt: 269 Schmelzpunkt: 114…118 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003347 CAS-Nummer: 25016-01-7 EINECS: 246-564-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...