3-Brom-4-hydroxybenzonitril ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A19400.06 A19400.14
A19400.06EA 46 EUR
A19400.06 A19400.14
3-Brom-4-hydroxybenzonitril ≥98%
3-Brom-4-hydroxybenzonitril

Purity: 98% CAS No.: 2315-86-8 Molecular Formula: C7H4BrNO Formula Weight: 198.02 Melting Point: 156-161° Beilstein Reference: 2207020 UN No.: 3439 MDL No.: MFCD00143096

Formel: C₇H₄BrNO
Siedepunkt: 406 °C (1013 hPa)
Schmelzpunkt: 156…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143096
CAS-Nummer: 2315-86-8
EINECS: 219-022-9
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR