D-Cycloserin

Lieferant: PanReac AppliChem
Achtung

Synonyme: D(+)-Cycloserin, D-4-Aminoisoxazolidin-3-on, (R)-(+)-Cycloserin

A1943.0001 A1943.0005 A1943.0025
A1943.0001EA 75.7 EUR
A1943.0001 A1943.0005 A1943.0025
D-Cycloserin
D-Cycloserin
Formel: C₃H₆N₂O₂
Molecular Weight: 102.09 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00005353
CAS-Nummer: 68-41-7
EINECS: 200-688-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR