Thiamin Hydrochlorid, (max. 5% H₂O) 99% (Trockengewicht)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Thiaminiumchlorid, Antiberiberifaktor Hydrochlorid, antineuritisches Vitamin Hydrochlorid, Aneurin Hydrochlorid, Thiaminchlorid Hydrochlorid, Vitamin B1 Hydrochlorid
A19560.0B
A19560.22
A19560.30
A19560.0BEA
390
EUR
A19560.0B
A19560.22
A19560.30
Thiamin Hydrochlorid, (max. 5% H₂O) 99% (Trockengewicht)
Thiamin Hydrochlorid
Purity: 99%
CAS No.: 67-03-8
Molecular Formula: C12H17ClN4OS.HCl
Formula Weight: 337.28
Physical Form: Cryst powder
Sensitivity: Light Sensitive and Hygroscopic
Appearance: White to pale yellow
Melting Point: ca 250° dec.
Beilstein Reference: 3851771
Merck Reference: 14,9295
MDL No.: MFCD00012780
Formel:
C₁₂H₁₈Cl₂N₄OS Schmelzpunkt: ∼ 250 °C (Zersetzung) Dichte: 1,401 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012780 CAS-Nummer: 67-03-8 EINECS: 200-641-8 Merck Index: 14,9295 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...