Thiamin Hydrochlorid, (max. 5% H₂O) 99% (Trockengewicht)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Thiaminiumchlorid, Antiberiberifaktor Hydrochlorid, antineuritisches Vitamin Hydrochlorid, Aneurin Hydrochlorid, Thiaminchlorid Hydrochlorid, Vitamin B1 Hydrochlorid

A19560.0B A19560.22 A19560.30
A19560.0BEA 390 EUR
A19560.0B A19560.22 A19560.30
Thiamin Hydrochlorid, (max. 5% H₂O) 99% (Trockengewicht)
Thiamin Hydrochlorid

Purity: 99% CAS No.: 67-03-8 Molecular Formula: C12H17ClN4OS.HCl Formula Weight: 337.28 Physical Form: Cryst powder Sensitivity: Light Sensitive and Hygroscopic Appearance: White to pale yellow Melting Point: ca 250° dec. Beilstein Reference: 3851771 Merck Reference: 14,9295 MDL No.: MFCD00012780

Formel: C₁₂H₁₈Cl₂N₄OS
Schmelzpunkt: ∼ 250 °C (Zersetzung)
Dichte: 1,401 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012780
CAS-Nummer: 67-03-8
EINECS: 200-641-8
Merck Index: 14,9295

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR