3-(Trifluormethoxy)anilin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98%
CAS No.: 1535-73-5
Molecular Formula: C7H6F3NO
Formula Weight: 177.13
Flash Point: 85°(185°F)
Refractive Index: 1.4660
Odour: Characteristic
Boiling Point: 72-75°/8mm
Beilstein Reference: 776921
UN No.: 2810
MDL No.: MFCD00041511
Formel:
C₇H₆F₃NO Siedepunkt: 72…75 °C (8 mmHg) Dichte: 1,325 Flammpunkt: 85 °C (185 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041511 CAS-Nummer: 1535-73-5 EINECS: 216-256-3 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...