Trimethylsilylcyanid ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97%
CAS No.: 7677-24-9
Molecular Formula: C4H9NSi
Formula Weight: 99.21
Flash Point: 1°(33°F)
Physical Form: Liquid
Density: 0.783
Refractive Index: 1.3910
Sensitivity: Moisture Sensitive
Odour: Almond-like
Melting Point: 11-12°
Boiling Point: 118-119°
Beilstein Reference: 1737612
UN No.: 3383
MDL No.: MFCD00001765
Formel:
C₄H₉NSi Siedepunkt: 118…119 °C Schmelzpunkt: 11…12 °C Dichte: 0,783 Flammpunkt: 1 °C (33 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001765 CAS-Nummer: 7677-24-9 EINECS: 231-657-3 UN: 3383 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...