2,4-Dichlor-5-methylbenzolsulfonylchlorid ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2,4-Dichlor-5-methylbenzolsulfochlorid, 2,4-Dichlor-5-methylbenzolsulfonsäurechlorid, 2,4-Dichlor-5-methylbenzol-1-sulfonylchlorid

A19718.09 A19718.18 A19718.88
A19718.09EA 161 EUR
A19718.09 A19718.18 A19718.88
2,4-Dichlor-5-methylbenzolsulfonylchlorid ≥97%
2,4-Dichlor-5-methylbenzolsulfonylchlorid

Purity: 97% CAS No.: 28286-86-4 Molecular Formula: C7H5Cl3O2S Formula Weight: 259.54 Melting Point: 70-73° UN No.: 3261 MDL No.: MFCD00044016

Formel: C₇H₅Cl₃O₂S
Molecular Weight: 259.54
Schmelzpunkt: 70…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00044016
CAS-Nummer: 28286-86-4
EINECS: 248-938-1
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR