4-(Trifluormethoxy)anisol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A19746.06
A19746.14
A19746.06EA
26.5
EUR
A19746.06
A19746.14
4-(Trifluormethoxy)anisol ≥98%
4-(Trifluormethoxy)anisol
Purity: 98+%
CAS No.: 710-18-9
Molecular Formula: C8H7F3O2
Formula Weight: 192.14
Flash Point: 58°(136°F)
Refractive Index: 1.4320
Boiling Point: 163-165°
Beilstein Reference: 2556809
UN No.: 1993
MDL No.: MFCD00216942
Formel:
C₈H₇F₃O₂ Siedepunkt: 163…165 °C Dichte: 1,272 Flammpunkt: 58 °C (136 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216942 CAS-Nummer: 710-18-9 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...