2,8-Bis(trifluormethyl)-4-chinolinol 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,8-Bis(trifluoromethyl)-4-hydroxyquinoline, 2,8-Bis(trifluoromethyl)-4-quinolinol

A19750.06
A19750.06EA 179 EUR
A19750.06
2,8-Bis(trifluormethyl)-4-chinolinol 97%
2,8-Bis(trifluoromethyl)quinolin-4-ol

Purity: 97% CAS No.: 35853-41-9 Molecular Formula: C11H5F6NO Formula Weight: 281.16 Melting Point: 129-135° MDL No.: MFCD00075091

Formel: C₁₁H₅F₆NO
Molecular Weight: 281,16 g/mol
Schmelzpunkt: 129…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075091
CAS-Nummer: 35853-41-9
EINECS: 252-762-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR