1-Brom-3-chlor-4-fluorbenzol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A19782.06
A19782.14
A19782.22
A19782.06EA
40.8
EUR
A19782.06
A19782.14
A19782.22
1-Brom-3-chlor-4-fluorbenzol ≥98%
1-Brom-3-chlor-4-fluorbenzol
Purity: 98+%
CAS No.: 60811-21-4
Molecular Formula: C6H3BrClF
Formula Weight: 209.45
Flash Point: 88°(190°F)
Refractive Index: 1.5540
Boiling Point: 188-190°
Beilstein Reference: 2206652
UN No.: 3082
MDL No.: MFCD00051794
Formel:
C₆H₃BrClF Siedepunkt: 188…190 °C Dichte: 1,727 Flammpunkt: 88 °C (190 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051794 CAS-Nummer: 60811-21-4 EINECS: 405-580-2 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...