N-Methylmorpholin-N-oxid 50% (w/w) in wässriger Lösung
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A19802.0B
A19802.22
A19802.30
A19802.0BEA
167
EUR
A19802.0B
A19802.22
A19802.30
N-Methylmorpholin-N-oxid 50% (w/w) in wässriger Lösung
N-Methylmorpholin-N-oxid
CAS No.: 7529-22-8
Molecular Formula: C5H11NO2
Formula Weight: 117.15
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Density: 1.140
Refractive Index: 1.4220
Appearance: Colourless to yellow
Melting Point: -20°
Boiling Point: 118-119°
MDL No.: MFCD00005947
Formel:
C₅H₁₁NO₂ Siedepunkt: 118…119 °C Schmelzpunkt: –20 °C Dichte: 1,140 Lagertemperatur: Kühlschrank |
MDL:
MFCD00005947 CAS-Nummer: 7529-22-8 EINECS: 231-391-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...