4-Ethoxybenzonitril ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: p-Ethoxybenzonitrile

A19806.06 A19806.14 A19806.22
A19806.06EA 39.3 EUR
A19806.06 A19806.14 A19806.22
4-Ethoxybenzonitril ≥98%
4-Ethoxybenzonitril

Purity: 98% CAS No.: 25117-74-2 Molecular Formula: C9H9NO Formula Weight: 147.18 Melting Point: 60-63° Boiling Point: 258° UN No.: 3439 MDL No.: MFCD00001819

Formel: C₉H₉NO
Molecular Weight: 147.18 g/mol
Siedepunkt: 258 °C
Schmelzpunkt: 60…63 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001819
CAS-Nummer: 25117-74-2
UN: 3439
ADR: 6.1 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR