Nerol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

A19808.AE
A19808.AEEA 61.4 EUR
A19808.AE
Nerol ≥97%
Nerol

Purity: 97% CAS No.: 106-25-2 Molecular Formula: C10H18O Formula Weight: 154.25 Flash Point: 77°(171°F) Refractive Index: 1.4750 Boiling Point: 103-105°/9mm Beilstein Reference: 1722455 Merck Reference: 14,6475 MDL No.: MFCD00063204

Formel: C₁₀H₁₈O
Siedepunkt: 103…105 °C (9 mmHg)
Dichte: 0,87
Flammpunkt: 77 °C (171 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00063204
CAS-Nummer: 106-25-2
EINECS: 203-378-7
Merck Index: 14,6475

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR