1,2-Dibrom-1-chlor-1,2,2-trifluorethan ≥98%

Lieferant: Thermo Fisher Scientific

A19856.14
A19856.14EA 27.6 EUR
A19856.14
1,2-Dibrom-1-chlor-1,2,2-trifluorethan ≥98%
1,2-Dibrom-1-chlor-1,2,2-trifluorethan
Formel: BrClFCCBrF₂
MDL: MFCD00039316
CAS-Nummer: 354-51-8
EINECS: 206-562-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR