Geranylacetat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 98%
CAS No.: 105-87-3
Molecular Formula: (CH3)2C=CHCH2CH2C(CH3)=CHCH2O2CCH3
Formula Weight: 196.29
Storage Temperature: 2°C to 8°C
Flash Point: 104°(219°F)
Physical Form: Liquid
Density: 0.91
Refractive Index: 1.461
Appearance: Colourless
Odour: Pleasant
Boiling Point: 137-139°/25mm
MDL No.: MFCD00015037
Formel:
C₁₂H₂₀O₂ Siedepunkt: 137…139 °C (25 mmHg) Dichte: 0,91 Flammpunkt: 104 °C (219 °F) |
MDL:
MFCD00015037 CAS-Nummer: 105-87-3 EINECS: 203-341-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...