1-Propanol ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99+%
CAS No.: 71-23-8
Molecular Formula: CH3CH2CH2OH
Formula Weight: 60.10
Flash Point: 15°(59°F)
Refractive Index: 1.3850
Odour: Alcohol-like
Melting Point: -127°
Boiling Point: 97°
Beilstein Reference: 1098242
Merck Reference: 14,7842
UN No.: 1274
MDL No.: MFCD00002941
Formel:
H₃CCH₂CH₂OH Siedepunkt: 97 °C Schmelzpunkt: –127 °C Dichte: 0,804 Flammpunkt: 15 °C (59 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002941 CAS-Nummer: 71-23-8 EINECS: 200-746-9 UN: 1274 ADR: 3,II Merck Index: 14,7842 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...