Sodium-3-bromopropanesulphonate 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A19930.06
A19930.22
A19930.06EA
32.3
EUR
A19930.06
A19930.22
Sodium-3-bromopropanesulphonate 97%
Sodium-3-bromopropanesulphonate
Purity: 97%
CAS No.: 55788-44-8
Molecular Formula: Br(CH2)3SO3Na
Formula Weight: 225.05
Melting Point: ca 256° dec.
MDL No.: MFCD00024999
Formel:
C₃H₇BrO₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024999 CAS-Nummer: 55788-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...