3-((Trifluormethyl)thio)benzoesäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A19975.03
A19975.06
A19975.03EA
52
EUR
A19975.03
A19975.06
3-((Trifluormethyl)thio)benzoesäure ≥97%
3-((Trifluormethyl)thio)benzoesäure
Purity: 97%
CAS No.: 946-65-6
Molecular Formula: C8H5F3O2S
Formula Weight: 222.18
Melting Point: 71-74°
MDL No.: MFCD00236337
Formel:
C₈H₅F₃O₂S Schmelzpunkt: 71…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236337 CAS-Nummer: 946-65-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...