3-((Trifluormethyl)thio)benzoesäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

A19975.03 A19975.06
A19975.03EA 52 EUR
A19975.03 A19975.06
3-((Trifluormethyl)thio)benzoesäure ≥97%
3-((Trifluormethyl)thio)benzoesäure

Purity: 97% CAS No.: 946-65-6 Molecular Formula: C8H5F3O2S Formula Weight: 222.18 Melting Point: 71-74° MDL No.: MFCD00236337

Formel: C₈H₅F₃O₂S
Schmelzpunkt: 71…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00236337
CAS-Nummer: 946-65-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR