3,5-Dichlorphenylisocyanat ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3,5-Dichlorophenylisocyantae
A19991.06
A19991.14
A19991.06EA
86.6
EUR
A19991.06
A19991.14
3,5-Dichlorphenylisocyanat ≥97%
3,5-Dichlorphenylisocyanat
Purity: 97%
CAS No.: 34893-92-0
Molecular Formula: C7H3Cl2NO
Formula Weight: 188.01
Flash Point: 134°(273°F)
Melting Point: 28-32°
Boiling Point: 242-244°
Beilstein Reference: 510282
UN No.: 2250
MDL No.: MFCD00013859
Formel:
C₇H₃Cl₂NO Molecular Weight: 188.01 g/mol Siedepunkt: 242…244 °C Schmelzpunkt: 28…32 °C Dichte: 1.380 Flammpunkt: 134 °C (273 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013859 CAS-Nummer: 34893-92-0 EINECS: 252-276-9 UN: 2250 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...