2,3-Dimethylanisol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A19992.09
A19992.18
A19992.09EA
52.4
EUR
A19992.09
A19992.18
2,3-Dimethylanisol ≥97%
2,3-Dimethylanisol
Purity: 97%
CAS No.: 2944-49-2
Molecular Formula: C9H12O
Formula Weight: 136.19
Flash Point: 67°(152°F)
Refractive Index: 1.5200
Melting Point: 29°
Boiling Point: 195°
Beilstein Reference: 1859736
MDL No.: MFCD00008376
Formel:
C₉H₁₂O Siedepunkt: 195 °C Schmelzpunkt: 29 °C Dichte: 0,984 Flammpunkt: 67 °C (152 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008376 CAS-Nummer: 2944-49-2 EINECS: 220-948-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...