3,6-Dichlor-4-methylpyridazin ≥97%

Lieferant: Thermo Scientific
Achtung

A19998.06
A19998.06EA 131 EUR
A19998.06
3,6-Dichlor-4-methylpyridazin ≥97%
3,6-Dichloro-4-methylpyridazine

Purity: 97% CAS No.: 19064-64-3 Molecular Formula: C5H4Cl2N2 Formula Weight: 163.01 Melting Point: 86-88° Boiling Point: 149-151°/21mm MDL No.: MFCD00006465

Formel: C₅H₄Cl₂N₂
Molecular Weight: 163,01 g/mol
Siedepunkt: 149…151 °C (21 mmHg)
Schmelzpunkt: 86…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006465
CAS-Nummer: 19064-64-3
EINECS: 242-794-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR