G418 Disulfat

Lieferant: PanReac AppliChem
Achtung

Synonyme: G-418 Sulfat, G418 Sulfat, Geneticin®, G-418 Disulfat

A2167.0001
A2167.0001EA 118 EUR
A2167.0001
G418 Disulfat
G418 disulphate
Formel: C₂₀H₄₄N₄O₁₈S₂
Molecular Weight: 692,72 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00058314
CAS-Nummer: 108321-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR