Luminol (3-Aminophthalhydrazid)

Lieferant: PanReac AppliChem

Synonyme: NSC 5064

A2185.0005
A2185.0005EA 77.7 EUR
A2185.0005
Luminol (3-Aminophthalhydrazid)
Luminol (3-Aminophthalhydrazid)
Formel: C₈H₇N₃O₂
Molecular Weight: 177.16 g/mol
MDL: MFCD00006890
CAS-Nummer: 521-31-3
EINECS: 208-309-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR