Lecithin ∼90% Soy Bean

Lieferant: PanReac AppliChem

A2218.0100
A2218.0100EA 220 EUR
A2218.0100
Lecithin ∼90% Soy Bean
Lecithin
MDL: MFCD00147406
CAS-Nummer: 8030-76-0
EINECS: 310-129-7
Merck Index: 12,05452

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR