XTT Natrium Salz

Lieferant: PanReac AppliChem

Synonyme: 2,3-Bis(2-methoxy-4-nitro-5-sulphophenyl)-2H-tetrazolium-5-carboxanilide, XTT sodium, Sodium 5,5'-(5-(phenylcarbamoyl)-2H-tetrazole-3-ium-2,3-diyl)bis(4-methoxy-2-nitrobenzenesulphonate), sodium 2,3,-bis(2-methoxy-4-nitro-5-sulphophenyl)-5-[(phenylamino)-carbonyl]-2H-tetrazolium inner salt

A2240.0100 A2240.0500
A2240.0100EA 263 EUR
A2240.0100 A2240.0500
XTT Natrium Salz
XTT sodium salt
Formel: C₂₂H₁₇N₇O₁₃S₂Na
Molecular Weight: 673.52 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00083517
CAS-Nummer: 111072-31-2

Bestellen

Spezifikation Testergebnisse

λmax. 283 - 287 nm
E 1 %/1 cm, 285 nm 14,800 - 16,800


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR