Dextran, M.W. ~ 40,000
Lieferant: PanReac AppliChem
A2249.0500
A2249.0500EA
197
EUR
A2249.0500
Dextran, M.W. ~ 40,000
Dextran
Formel:
(C₆H₁₀O₅)n |
MDL:
MFCD00130935 CAS-Nummer: 9004-54-0 EINECS: 232-677-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...