di-Natrium-β-glycerophosphat Pentahydrat

Lieferant: PanReac AppliChem

A2253.0100 A2253.0500
A2253.0100EA 243 EUR
A2253.0100 A2253.0500
di-Natrium-β-glycerophosphat Pentahydrat
di-Natrium-β-glycerophosphat Pentahydrat
Formel: C₃H₇Na₂O₆P·5H₂O
MDL: MFCD00002135
CAS-Nummer: 13408-09-8
EINECS: 212-464-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR