Ethidiumbromid solution_SC 0,07 %

Lieferant: PanReac AppliChem

Synonyme: 3,8-Diamino-5-ethyl-6-phenylphenanthridiniumbromid

A2273.0005 A2273.0015
A2273.0005EA 30.3 EUR
A2273.0005 A2273.0015
Ethidiumbromid solution_SC 0,07 %
Ethidiumbromid

Ethidium bromide solutions with a concentration of 0,7 mg/ml.


Lieferumfang: Supplied in a dropper bottle.

Formel: C₂₁H₂₀BrN₃
Molecular Weight: 394.31 g/mol
Siedepunkt: 72
Lagertemperatur: Raumtemperatur
MDL: MFCD00011724
CAS-Nummer: 1239-45-8
Merck Index: 13,04751

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR