Heparin-Natriumsalz, fest
Lieferant: PanReac AppliChem
Synonyme:
Reviparin sodium
A3004.0005
A3004.0005EA
424
EUR
A3004.0005
Heparin-Natriumsalz, fest
Heparin sodium salt
MDL:
MFCD00081689 CAS-Nummer: 9041-08-1 |
Spezifikation Testergebnisse
State of Matter | Solid |
Origin | Porcine mucosa |
Storage | Store at Room Temperature |
Assay | Min. 150 I.U./mg |
Solubility (1%; H2O) | Clear, Colorless |
pH (1%; H2O; 20 °C) | 5,5 - 8,0 |
Heavy metals (as Pb) | Max. 0,003% |
Loss on drying | Max. 8% |
Sodium (Na) | 9,5 - 12,5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...