Azomethin H Natriumsalz Hydrat ~95%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: Natriumhydrogen-4-hydroxy-5-(salicylidenamino)naphthalin-2,7-disulfonat Hydrat, 8-Hydroxy-1-(salicylidenamino)naphthalen-3,6-disulfonsäure-6-natriumsalz Hydrat, Azomethine-H Mononatriumsalz Hydrat

M-Clarity™ Qualitätsstufe = MQ200

A3144-1G A3144-25G A3144-5G
A3144-1GEA 63.4 EUR
A3144-1G A3144-25G A3144-5G
Azomethin H Natriumsalz Hydrat ~95%, Sigma-Aldrich®
Azomethin H Natriumsalz Hydrat

Azomethine-H is used as a color development agent. It is used for the determination of boron, mainly in soil samples. Azomethine-H monosodium salt hydrate has been used for the colorimetric determination of boron in soil and plants.

Formel: C₁₇H₁₂NNaO₈S₂·xH₂O
Molecular Weight: 463.42 g/mol
Lagertemperatur: Room Temperature
MDL: MFCD00149588
CAS-Nummer: 206752-32-1
EINECS: 227-698-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=039fa8fabf130be6b193fa11648e1ec2d7bc445c&vid=3964aa9c3af9f4f9f45bfc0571c0424ef21f45c1&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form) Powder
Appearance (Colour) Faint Yellow to Dark Yellow and Faint Orange to Dark Orange
Solubility (Solvent) 1N NaOH
Solubility (Conc) 49.00 - 51.00 mg/mL
Solubility (Turbidity) Clear to Hazy
Solubility (Colour) Faint Yellow to Dark Yellow and Faint Orange to Dark Orange
Purity (TLC) ≥ 95.00%
NMR (Solvent) D2O
Identity by NMR Consistent
NMR (Residual Solvent 1) E/E
NMR (% Solvent 1) ≤ 1.00%
NMR (Residual Solvent 2) Acetone
NMR (% Solvent 2) ≤ 0.50%
UV (Solvent) Water
UV (Lambda max 1) 235 - 239 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR