Dextransulfat Natriumsalz, M.W. ~ 40,000

Lieferant: PanReac AppliChem

Synonyme: Dextran sulphate sodium from Dextran

A3261.0250
A3261.0250EA 624 EUR
A3261.0250
Dextransulfat Natriumsalz, M.W. ~ 40,000
Dextran sulphate sodium salt
Formel: (C₆H₇Na₃O₁₄S₃)n
MDL: MFCD00081551
CAS-Nummer: 9011-18-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR