Dithiothreitol (DTT, Cleland's Reagenz) 1 mol/l aqueous solution

Lieferant: PanReac AppliChem

Synonyme: DTT, (R*,R*)-1,4-Dimercapto-2,3-butandiol

A3668.0100 A3668.0050
A3668.0100EA 75.9 EUR
A3668.0100 A3668.0050
Dithiothreitol (DTT, Cleland's Reagenz) 1 mol/l aqueous solution
Dithiothreitol (DTT, Cleland's Reagenz)
Formel: C₄H₁₀O₂S₂
Molecular Weight: 154.25 g/mol
MDL: MFCD00004877
CAS-Nummer: 3483-12-3
EINECS: 222-468-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR