L(+)-Cystein
Lieferant: PanReac AppliChem
|
Achtung
|
Synonyme:
(R)-(+)-2-Amino-3-mercaptopropansäure, H-Cys-OH, (R)-(+)-Cystein
A3694.0100
A3694.0100EA
164
EUR
A3694.0100
L(+)-Cystein
L(+)-Cystein
Formel:
HSCH₂CH(NH₂)CO₂H Molecular Weight: 121,16 g/mol Siedepunkt: 254 °C (1013 hPa) Schmelzpunkt: 115 °C Dichte: 1,523 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064306 CAS-Nummer: 52-90-4 EINECS: 200-158-2 Merck Index: 13,02810 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...