L(+)-Glutamin
Lieferant: PanReac AppliChem
Synonyme:
L(+)-Glutaminsäure-5-amid, Q, (S)-(+)-2-Amino-1,5-pentandisäure-5-amid, (S)-(+)-Glutamin, L(+)-2-Amino-1,5-pentandisäure-5-amid, H-Gln-OH, (S)-(+)-Glutaminsäure-5-amid, Gln, Levoglutamid
A3704.1000
A3704.1000EA
337
EUR
A3704.1000
L(+)-Glutamin
L(+)-Glutamin
Formel:
C₅H₁₀N₂O₃ Molecular Weight: 146,15 g/mol Siedepunkt: 355 °C (1013 hPa) Schmelzpunkt: 185…186 °C Dichte: 1,5326 g/cm³ (130 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008044 CAS-Nummer: 56-85-9 EINECS: 200-292-1 Merck Index: 13,04484 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...