L(+)-Histidin Base, BioChemica®

Lieferant: PanReac AppliChem

A3738.0100 A3738.1000
A3738.0100EA 46 EUR
A3738.0100 A3738.1000
L(+)-Histidin Base, BioChemica®
L(+)-Histidin

  • All amino acids from AppliChem are of non-animal origin!

Formel: C₆H₉N₃O₂
Schmelzpunkt: 272…273 °C
Dichte: 1,44 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064315
CAS-Nummer: 71-00-1
EINECS: 200-745-3
Merck Index: 13,04741
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=badfdb36aac0611e5823a7d05e6cba944f6650fa&vid=9e9f9d881a5b3314c7f342312e77e13493901200&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (titr.) min. 99 %
α20°C/D; 11 %, HCl 120 g/l +11.8° - +12.8°
Heavy metals (as Pb) max. 0.001 %
Insoluble matter passes test
Other amino acids max. 0.5 %
Ammonium max. 0.1 %
Chloride max. 0.02 %
Sulfate max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR