Acetyl-coenzyme A
Lieferant: PanReac AppliChem
A3753.0100
A3753.0100EA
366
EUR
A3753.0100
Acetyl-coenzyme A
Enzyme
- Assay (enzymatic): min. 83%
- Assay (HPLC): approx. 95%
- Loss on drying: max. 5%
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...