Buffer, triethylammonium acetate, 1 M solution, pH 7,0
Lieferant: PanReac AppliChem
A3846.0500
A3846.1000
A3846.0500EA
91.4
EUR
A3846.0500
A3846.1000
Buffer, triethylammonium acetate, 1 M solution, pH 7,0
Puffer
Biologische Puffer
Used for the purification of oligonucleotides.
CAS No.: 5204-74-0
Molecular Formula: C₆H₁₅N · CH₃COOH
Formula Weight: 161,24 g/mol
Density: 1,001 (at 20 °C)
Physical Form: Liquid
Non-volatile Matter: Max. 0,01%
Storage Temperature: Ambient
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...