1-Amino-9,10-anthrachinon 97%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 1-Aminoanthrachinon

MQ100

A39009-100G
A39009-100GEA 54.6 EUR
A39009-100G
1-Amino-9,10-anthrachinon 97%, Sigma-Aldrich®
1-Amino-9,10-anthrachinon

1-Aminoanthraquinone has the ability to increase the solubility of derivatives of anthraquinone in supercritical carbon dioxide.

Formel: C₁₄H₉NO₂
Molecular Weight: 223.23 g/mol
Siedepunkt: 465
Schmelzpunkt: 251…254 °C
Lagertemperatur: Room Temperature
MDL: MFCD00001213
CAS-Nummer: 82-45-1
EINECS: 201-423-5
Merck Index: 13,00416
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=728ad5bf6413a649184f49ad2f2234816898df48&vid=63366f56aaaea3b17c8997350394aef337d5bf45&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form) Powder or Crystal or Chunk(s)
Infrared spectrum Conforms to Structure
Carbon 72.7 - 78.0 %
Nitrogen 6.0 - 6.6 %
Purity (HPLC) ≥96.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR