Azur B (Azur I) 89 Dye, certified_Biological Stain Commission, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: C.I. 52010, N,N,N′-Trimethylthionin, Azure B, Trimethylthionine chloride, 3-(Dimethylamino)-7-(methylamino)phenothiazin-5-ium chloride, Methylene Azur B

MQ200

A4043-25G A4043-5G
A4043-25GEA 605 EUR
A4043-25G A4043-5G
Azur B (Azur I) 89 Dye, certified_Biological Stain Commission, Sigma-Aldrich®
Azur B (Azur I)
Formel: C₁₅H₁₆ClN₃S
Molecular Weight: 305.83 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011935
CAS-Nummer: 531-55-5
EINECS: 208-511-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=600ef5cf3e062a4a587e204a7525059448b68bb0&vid=8472fabd8cb28ce018854ce4ca2ef091bb7e36ab&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR