TRIS Pufferlösung pH 8 (1 mol/l)

Lieferant: PanReac AppliChem

A4577.1000
A4577.1000EA 42.9 EUR
A4577.1000
TRIS Pufferlösung pH 8 (1 mol/l)
Puffer Klonpuffer

  • DNases/RNases/Proteases: not detectable
  • pH (20°C; adjusted with HCl): 8.0 ± 0.1
  • Composition: Tris: 121.14 g/L (1 M)

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR