Mupirocin USP, pharmazeutischer Qualität
Lieferant: PanReac AppliChem
Synonyme:
9-(((E)-4-((2S,3R,4R,5S)-3,4-Dihydroxy-5-(((2S,3S)-3-((2S,3S)-3-hydroxybutan-2-yl)oxiran-2-yl)methyl)tetrahydro-2H-pyran-2-yl)-3-methylbut-2-enoyl)oxy)nonanoic acid
A4718.0025
A4718.0025EA
452
EUR
A4718.0025
Mupirocin USP, pharmazeutischer Qualität
Mupirocin
Standards: Meets the requirements of the US Pharmacopoeia (USP 40).
Formel:
C₂₆H₄₄O₉ Molecular Weight: 500.629 g/mol |
MDL:
MFCD01711620 CAS-Nummer: 12650-69-0 |
Spezifikation Testergebnisse
Form | Solid |
Assay (HPLC, calculated on dried substances) | 920 - 1020 µg/mg |
Identity | Passes test |
Crystallinity | Passes test |
pH (saturated solution; H₂O; 25 °C) | 3,5 - 4,5 |
Water (K.F.) | Max. 1,0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...