Acrylamid-Bisacrylamid (29:1) 30% in wässriger Lösung

Lieferant: PanReac AppliChem
Gefahr

A4983.0500
A4983.0500EA 77.6 EUR
A4983.0500
Acrylamid-Bisacrylamid (29:1) 30% in wässriger Lösung
Acrylamid-Bis (32:1)
MDL: MFCD00080848
UN: 3426
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR