Ammoniumsulfat 99,0, reagent

Lieferant: Honeywell Chemicals

Synonyme: Diammoniumsulfat, di-Ammoniumsulfat

A5132-1KG A5132-5KG
A5132-1KGEA 52.19 EUR
A5132-1KG A5132-5KG
Ammoniumsulfat 99,0, reagent
Ammoniumsulfat

  • ≥95% for general laboratory use.

Formel: (NH₄)₂SO₄
Molecular Weight: 132.14 g/mol
Schmelzpunkt: 350 °C
Dichte: 1.760 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003391
CAS-Nummer: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Bestellen

Spezifikation Testergebnisse

Assay 99.0 - 101.0 %
Colour of the solution complying
Clearness of the solution complying
Iron (Fe) Max. 10 ppm
Lead (Pb) Max. 3 ppm
Selenium (Se) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR